Product Details
- SNP ID
-
rs17156046
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:1179459 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGCAAAAACAAAACCCACAGAAA[C/T]CCTAACTCTATCCCTTGATATCACA
- Phenotype
-
MIM: 602065
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADARB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76048130] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADARB2
- Gene Name
- adenosine deaminase, RNA specific B2 (inactive)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018702.3 |
6280 |
UTR 3 |
|
|
NP_061172.1 |
View Full Product Details