Product Details

SNP ID
rs17151353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:134033463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATGAAAGCAAAAACTGGAAGATA[C/T]CATAACACTGAATTAAGAAGCAGAC
Phenotype
MIM: 601541
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRD3 PubMed Links

Gene Details

Gene
BRD3
Gene Name
bromodomain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007371.3 2847 UTR 3 NP_031397.1
XM_006717291.2 2847 UTR 3 XP_006717354.1
XM_011519052.2 2847 UTR 3 XP_011517354.1
XM_017015165.1 2847 UTR 3 XP_016870654.1
XM_017015166.1 2847 UTR 3 XP_016870655.1
Gene
LINC00094
Gene Name
long intergenic non-protein coding RNA 94
There are no transcripts associated with this gene.

View Full Product Details