Product Details

SNP ID
rs17853670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118825884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCCAACTTTCTGGAGTTAATC[A/G]GAATGGTAAGGGAGGAAGAGGATTG
Phenotype
MIM: 300701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC12 PubMed Links

Gene Details

Gene
ZCCHC12
Gene Name
zinc finger CCHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312891.1 1142 Missense Mutation AGA,GGA R214G NP_001299820.1
NM_173798.3 1142 Missense Mutation AGA,GGA R214G NP_776159.1

View Full Product Details