Product Details

SNP ID
rs16849238
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:165667062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGACATCTAACCCAATGCTTTACA[C/T]AGATCAGAATCTTAAGTGTTTATTA
Phenotype
MIM: 602733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH9A1 PubMed Links

Gene Details

Gene
ALDH9A1
Gene Name
aldehyde dehydrogenase 9 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000696.3 Intron NP_000687.3
XM_011509294.2 Intron XP_011507596.1

View Full Product Details