Product Details

SNP ID
rs16916652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94249821 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTTGATGGAGCAATAATAGTCA[C/T]GATTCTGATGAGCCTAGTTCTACAA
Phenotype
MIM: 600164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEM PubMed Links

Gene Details

Gene
GEM
Gene Name
GTP binding protein overexpressed in skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005261.3 1823 UTR 3 NP_005252.1
NM_181702.2 1823 UTR 3 NP_859053.1
XM_017013315.1 1823 UTR 3 XP_016868804.1

View Full Product Details