Product Details
- SNP ID
-
rs17016977
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:121727390 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGTCTTTAATTGCTGGCGACAGA[A/G]AAGGCTGCAGTTTAGTACTTAAACC
- Phenotype
-
MIM: 611970
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NIFK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116084393] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NIFK
- Gene Name
- nucleolar protein interacting with the FHA domain of MKI67
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032390.4 |
1293 |
UTR 3 |
|
|
NP_115766.3 |
- Gene
- NIFK-AS1
- Gene Name
- NIFK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details