Product Details

SNP ID
rs16939906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85901900 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACTCGTTTGCCCAAAGAGGTCA[C/T]ATTTTAATCAGAAGAACAAAAATGC
Phenotype
MIM: 601565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF8 PubMed Links

Gene Details

Gene
IRF8
Gene Name
interferon regulatory factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002163.2 Intron NP_002154.1
XM_017023199.1 Intron XP_016878688.1

View Full Product Details