Product Details

SNP ID
rs17303157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72221670 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGCCCAACCATACATGTGCCTTC[A/G]GAATAACTGAAACAAGTATTTGCCT
Phenotype
MIM: 611989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS27 PubMed Links

Gene Details

Gene
MRPS27
Gene Name
mitochondrial ribosomal protein S27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286748.1 Intron NP_001273677.1
NM_001286751.1 Intron NP_001273680.1
NM_015084.2 Intron NP_055899.2

View Full Product Details