Product Details
- SNP ID
-
rs17851135
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:72968557 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATATGAAACATGGACAGATATC[A/G]ACAAAAGAACTAGCAGATTTTGTAA
- Phenotype
-
MIM: 613438
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FCHO2
PubMed Links
Gene Details
- Gene
- FCHO2
- Gene Name
- FCH domain only 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146032.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
NP_001139504.1 |
NM_138782.2 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
NP_620137.2 |
XM_017009016.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864505.1 |
XM_017009017.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864506.1 |
XM_017009018.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864507.1 |
XM_017009019.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864508.1 |
XM_017009020.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864509.1 |
XM_017009021.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864510.1 |
XM_017009022.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864511.1 |
XM_017009023.1 |
203 |
Silent Mutation |
TCA,TCG |
S31S |
XP_016864512.1 |
View Full Product Details