Product Details

SNP ID
rs17267170
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84059596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTTCATAGGAAAGATAAAAGTAA[C/G]CTAATCAGTCACATTTTGACAATTT
Phenotype
MIM: 300303
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 6955 UTR 3 NP_055311.1
XM_011530917.2 6955 Intron XP_011529219.1
XM_011530918.2 6955 Intron XP_011529220.1
XM_011530919.2 6955 Intron XP_011529221.1
XM_011530920.2 6955 Intron XP_011529222.1
XM_017029423.1 6955 Intron XP_016884912.1
XM_017029424.1 6955 Intron XP_016884913.1
XM_017029425.1 6955 Intron XP_016884914.1
XM_017029426.1 6955 Intron XP_016884915.1

View Full Product Details