Product Details

SNP ID
rs17020870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105249820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGGGAGCACCACAGAGTATGCG[C/T]GGGGTTCAGGCCAGGGCACCATAAT
Phenotype
MIM: 604838 MIM: 606237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR45 PubMed Links

Gene Details

Gene
GPR45
Gene Name
G protein-coupled receptor 45
There are no transcripts associated with this gene.

Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142621.2 3063 Intron NP_001136093.1
NM_004257.5 3063 Intron NP_004248.2
XM_011512193.2 3063 UTR 3 XP_011510495.1
XM_017005339.1 3063 Intron XP_016860828.1

View Full Product Details