Product Details
- SNP ID
-
rs16876647
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:47860612 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCACTACTGCCCATTCTTCACAATG[C/T]CTATCTTCCTTTATGATTTTTTTTC
- Phenotype
-
MIM: 616908
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTCHD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115875453] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTCHD4
- Gene Name
- patched domain containing 4
View Full Product Details