Product Details

SNP ID
rs17805348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49478520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCATTATTTTCATTTTTTAGTT[G/T]TCAGTTCTTACTAGATTACAGCTAT
Phenotype
MIM: 611506
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPQ PubMed Links

Gene Details

Gene
CENPQ
Gene Name
centromere protein Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018132.3 Intron NP_060602.2
XM_005249205.1 Intron XP_005249262.1
XM_011514723.2 Intron XP_011513025.2

View Full Product Details