Product Details

SNP ID
rs34820962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16622915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCCAGTATGGGTCCTCTGATG[C/T]ACAATAAGGTATGATTTAGTCCTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF624 PubMed Links

Gene Details

Gene
ZNF624
Gene Name
zinc finger protein 624
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020787.3 2063 Silent Mutation GTA,GTG V657V NP_065838.2
XM_006721562.3 2063 Silent Mutation GTA,GTG V657V XP_006721625.1
XM_011523970.2 2063 Silent Mutation GTA,GTG V655V XP_011522272.1

View Full Product Details