Product Details

SNP ID
rs17050736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139291216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATAACTAGGTTTTTTACCAGGAA[C/T]TGTCCAACTGGGTATTAGTTGAGTT
Phenotype
MIM: 603844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFC1 PubMed Links

Gene Details

Gene
NDUFC1
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184986.1 Intron NP_001171915.1
NM_001184987.1 Intron NP_001171916.1
NM_001184988.1 Intron NP_001171917.1
NM_001184989.1 Intron NP_001171918.1
NM_001184990.1 Intron NP_001171919.1
NM_001184991.1 Intron NP_001171920.1
NM_002494.3 Intron NP_002485.1

View Full Product Details