Product Details

SNP ID
rs17086394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56182624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGGGTAGGAGTTGCACACATAC[A/G]ACGTTGCAGTCCCTGTCCAAATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1211 PubMed Links

Gene Details

Gene
KIAA1211
Gene Name
KIAA1211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020722.1 Intron NP_065773.1
XM_005265752.2 Intron XP_005265809.1
XM_005265755.2 Intron XP_005265812.1
XM_005265756.2 Intron XP_005265813.1
XM_006714045.2 Intron XP_006714108.1
XM_011534397.2 Intron XP_011532699.1
XM_017008473.1 Intron XP_016863962.1
XM_017008474.1 Intron XP_016863963.1

View Full Product Details