Product Details

SNP ID
rs17104423
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:48520611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTAAGTTTATAATAAAAAAGGA[A/G]ATGTTACTTGTAGAAATTGGTCTTC
Phenotype
MIM: 616476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGBL4 PubMed Links

Gene Details

Gene
AGBL4
Gene Name
ATP/GTP binding protein like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323573.1 3661 Intron NP_001310502.1
NM_001323574.1 3661 Intron NP_001310503.1
NM_001323575.1 3661 Intron NP_001310504.1
NM_032785.3 3661 Intron NP_116174.3
XM_011542308.2 3661 Intron XP_011540610.1
XM_011542310.2 3661 Intron XP_011540612.1
XM_017002595.1 3661 UTR 3 XP_016858084.1
XM_017002596.1 3661 Intron XP_016858085.1
XM_017002597.1 3661 Intron XP_016858086.1
XM_017002598.1 3661 Intron XP_016858087.1

View Full Product Details