Product Details
- SNP ID
-
rs17468972
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
57
- Location
-
Chr.1:48102728 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTCACACATGTATCATCCAGCCTC[C/T]GAGGTGCCTCTGACAACCATAATTG
- Phenotype
-
MIM: 616392
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SKINT1L
PubMed Links
Gene Details
- Gene
- SKINT1L
- Gene Name
- Skint1 like (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details