Product Details

SNP ID
rs17063210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133960151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAATTGCTTGTGGGTTGACCTC[A/C]TTCCAGGATCTTTGCTCGTTACAGA
Phenotype
MIM: 605521
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TBPL1 PubMed Links

Gene Details

Gene
TBPL1
Gene Name
TATA-box binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253676.1 Intron NP_001240605.1
NM_004865.3 Intron NP_004856.1
XM_017011513.1 Intron XP_016867002.1

View Full Product Details