Product Details
- SNP ID
-
rs17061686
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132730151 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGAATAAAAAGGACAGATTAATG[A/G]CCTACTGGTTTAGACCTTTTCTTTT
- Phenotype
-
MIM: 606592
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
VNN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs540652229] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VNN3
- Gene Name
- vanin 3
View Full Product Details