Product Details

SNP ID
rs17291062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:114239327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTTAGAAGTCGTGAGTGATGAG[C/T]GGCACTTCTGTTTTCCACGTAGTAT
Phenotype
MIM: 603461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC16 PubMed Links

Gene Details

Gene
CDC16
Gene Name
cell division cycle 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005266206.1 Intron XP_005266263.1
XM_006719992.1 Intron XP_006720055.1
XM_006719993.1 Intron XP_006720056.1
XM_011534851.1 Intron XP_011533153.1
XM_011534853.1 Intron XP_011533155.1
XM_017020830.1 Intron XP_016876319.1
XM_017020831.1 Intron XP_016876320.1
XM_017020832.1 Intron XP_016876321.1
XM_017020833.1 Intron XP_016876322.1
Gene
MIR548AR
Gene Name
microRNA 548ar
There are no transcripts associated with this gene.

View Full Product Details