Product Details

SNP ID
rs17854475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33027310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGTGCGCAAGCTTTCCAAGATC[A/G]CCACGCTGCTGCTGGCGCGCAACTA
Phenotype
MIM: 606386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLIG2 PubMed Links

Gene Details

Gene
OLIG2
Gene Name
oligodendrocyte lineage transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005806.3 836 Missense Mutation ACC,GCC T150A NP_005797.1
XM_005260908.1 836 Missense Mutation ACC,GCC T150A XP_005260965.1

View Full Product Details