Product Details

SNP ID
rs16982501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:21132600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCTAGTTGGAGTACTCTTACC[A/G]TTCTCTTATTTTTAGTGTGTAATAA
Phenotype
MIM: 615757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIZ PubMed Links

Gene Details

Gene
KIZ
Gene Name
kizuna centrosomal protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163022.1 Intron NP_001156494.1
NM_001163023.1 Intron NP_001156495.1
NM_001276389.1 Intron NP_001263318.1
NM_018474.4 Intron NP_060944.3
XM_011529296.2 Intron XP_011527598.1
XM_011529297.2 Intron XP_011527599.1
XM_011529298.2 Intron XP_011527600.1
XM_011529299.2 Intron XP_011527601.1
XM_017027951.1 Intron XP_016883440.1
XM_017027952.1 Intron XP_016883441.1
XM_017027953.1 Intron XP_016883442.1
XM_017027954.1 Intron XP_016883443.1

View Full Product Details