Product Details

SNP ID
rs17370078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:19760014 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAACTCTGGCTTCAGATGTAGTCA[C/T]GTTCACTGAATGCAGAGCTTTTATT
Phenotype
MIM: 610222
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987276 PubMed Links

Gene Details

Gene
LOC107987276
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017028183.1 Intron XP_016883672.1
Gene
RIN2
Gene Name
Ras and Rab interactor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242581.1 Intron NP_001229510.1
NM_018993.3 Intron NP_061866.1
XM_005260731.2 Intron XP_005260788.1
XM_005260733.2 Intron XP_005260790.1
XM_006723574.3 Intron XP_006723637.1
XM_006723575.3 Intron XP_006723638.1
XM_006723577.2 Intron XP_006723640.1
XM_011529255.2 Intron XP_011527557.2
XM_011529257.1 Intron XP_011527559.1
XM_011529258.2 Intron XP_011527560.1
XM_011529259.2 Intron XP_011527561.1
XM_017027887.1 Intron XP_016883376.1
XM_017027888.1 Intron XP_016883377.1
XM_017027889.1 Intron XP_016883378.1
XM_017027890.1 Intron XP_016883379.1
XM_017027891.1 Intron XP_016883380.1
XM_017027892.1 Intron XP_016883381.1
XM_017027893.1 Intron XP_016883382.1

View Full Product Details