Product Details

SNP ID
rs16999434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17579952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATCTGTTATCTCTACCCAACTG[A/G]ATATATCTTTAGAATAGTTCAAAGT
Phenotype
MIM: 609114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSTN PubMed Links

Gene Details

Gene
DSTN
Gene Name
destrin, actin depolymerizing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011546.1 Intron NP_001011546.1
NM_006870.3 Intron NP_006861.1
XM_011529142.1 Intron XP_011527444.1
XM_011529143.1 Intron XP_011527445.1
XM_011529144.1 Intron XP_011527446.1

View Full Product Details