Product Details
- SNP ID
-
rs17000600
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:134378628 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCACAGGAGCTTTTCTGAAGCTG[C/T]GTTAGAGTAATGTTCCCGTAGTTGT
- Phenotype
-
MIM: 300414
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PHF6
PubMed Links
Gene Details
- Gene
- PHF6
- Gene Name
- PHD finger protein 6
View Full Product Details