Product Details

SNP ID
rs17330661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135033423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACGCAGTATTCTGAGTTTGCAAC[C/T]GTCTCTCTGATGTGTGCCTTTTGTT
Phenotype
MIM: 300213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM127A PubMed Links

Gene Details

Gene
FAM127A
Gene Name
family with sequence similarity 127 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078171.1 Intron NP_001071639.1

View Full Product Details