Product Details

SNP ID
rs955955
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:151039067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGACTCAGCTGGTAGAAGTAGAG[A/G]CTAGAGCTAAAGGATCAGGATGGAC
Phenotype
MIM: 611275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159642.1 Intron NP_001153114.1
NM_138278.3 Intron NP_612122.2
XM_011509236.1 Intron XP_011507538.1
XM_017000420.1 Intron XP_016855909.1
Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
There are no transcripts associated with this gene.

Gene
PRUNE1
Gene Name
prune exopolyphosphatase
There are no transcripts associated with this gene.

View Full Product Details