Product Details
- SNP ID
-
rs2091219
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
60
- Location
-
Chr.1:151039584 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAGCATGGCATGTTGGGGAATTCA[A/G]AGTATGAGCATGACTATTTTAATAT
- Phenotype
-
MIM: 611275
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BNIPL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115422057] are located under a probe and SNP(s) [rs114960392] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BNIPL
- Gene Name
- BCL2 interacting protein like
- Gene
- C1orf56
- Gene Name
- chromosome 1 open reading frame 56
There are no transcripts associated with this gene.
- Gene
- PRUNE1
- Gene Name
- prune exopolyphosphatase
There are no transcripts associated with this gene.
View Full Product Details