Product Details

SNP ID
rs7881612
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:171091514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAGGAACAGAAAAAAGTTAGGA[A/G]ACGTAATTTTCTTAGAGGGGAATGG
Phenotype
MIM: 136132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FMO3 PubMed Links

Gene Details

Gene
FMO3
Gene Name
flavin containing monooxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002294.2 Intron NP_001002294.1
NM_001319173.1 Intron NP_001306102.1
NM_001319174.1 Intron NP_001306103.1
NM_006894.5 Intron NP_008825.4
XM_011509345.2 Intron XP_011507647.1

View Full Product Details