Product Details

SNP ID
rs11542012
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:151983227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATTGCATGCAATGGTGAGGCCC[A/G]CAATTAGGGAAAAGAAGCTCTGGAA
Phenotype
MIM: 114085
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 738 Missense Mutation GCG,GTG A77V NP_002957.1

View Full Product Details