Product Details

SNP ID
rs11555721
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153638352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAGTGAATATTCGGGCTTCGAT[G/T]CAGCAACAACAGCAGCTAGCCAGTG
Phenotype
MIM: 614206 MIM: 176940 MIM: 601989
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHTOP PubMed Links

Gene Details

Gene
CHTOP
Gene Name
chromatin target of PRMT1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206612.1 502 Missense Mutation ATG,ATT M41I NP_001193541.1
NM_001244664.1 502 Missense Mutation ATG,ATT M41I NP_001231593.1
NM_001317077.1 502 Missense Mutation ATG,ATT M41I NP_001304006.1
NM_015607.3 502 Missense Mutation ATG,ATT M41I NP_056422.2
XM_017000993.1 502 Missense Mutation ATG,ATT M41I XP_016856482.1
Gene
S100A1
Gene Name
S100 calcium binding protein A1
There are no transcripts associated with this gene.

Gene
S100A13
Gene Name
S100 calcium binding protein A13
There are no transcripts associated with this gene.

View Full Product Details