Product Details

SNP ID
rs11556418
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153658778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTTCCGCCGCTGTATCGGGGG[C/T]AGGGACCCCGGTGGCGGGGCCCACA
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
There are no transcripts associated with this gene.

Gene
SNAPIN
Gene Name
SNAP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012437.5 125 Missense Mutation GCA,GTA A12V NP_036569.1

View Full Product Details