Product Details
- SNP ID
-
rs10801138
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:192640515 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTAAGGTCATCTAGAACCTAACT[A/G]TGCACACAAACATATCCCTTTAATA
- Phenotype
-
MIM: 607190
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RGS13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112587334,rs76612809] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RGS13
- Gene Name
- regulator of G-protein signaling 13
View Full Product Details