Product Details

SNP ID
rs6671688
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:155021789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTACTGAGGCCATCACGAACTTC[C/T]CCTAGACTAAGACAAGCTTCTCTCT
Phenotype
MIM: 607646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCST2 PubMed Links

Gene Details

Gene
DCST2
Gene Name
DC-STAMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144622.2 Intron NP_653223.2
XM_011509188.2 Intron XP_011507490.1
XM_011509189.2 Intron XP_011507491.1
Gene
ZBTB7B
Gene Name
zinc finger and BTB domain containing 7B
There are no transcripts associated with this gene.

View Full Product Details