Product Details
- SNP ID
-
hCV42508927
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:158291174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTTCCATGTCATCCAGATCTT[C/T]TCATTTGTCAACCAATCCTGGGCAC
- Phenotype
-
MIM: 188360
MIM: 188340
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD1B
PubMed Links
Gene Details
- Gene
- CD1B
- Gene Name
- CD1b molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001764.2 |
394 |
Intron |
|
|
NP_001755.1 |
- Gene
- CD1C
- Gene Name
- CD1c molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001765.2 |
394 |
Silent Mutation |
TTC,TTT |
F34F |
NP_001756.2 |
View Full Product Details