Product Details

SNP ID
rs11554223
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32222570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTACTGCAGGGCCATGAGCGGTC[C/G]ATTACGCAGATTAAGTATAACCGCG
Phenotype
MIM: 603911
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCDC2B PubMed Links

Gene Details

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003757.3 569 Silent Mutation TCC,TCG S12S NP_003748.1
XM_017002671.1 569 UTR 5 XP_016858160.1
Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 569 Intron NP_061991.3
XM_011541795.2 569 Intron XP_011540097.1
XM_017001812.1 569 Intron XP_016857301.1
XM_017001813.1 569 Intron XP_016857302.1
XM_017001814.1 569 Intron XP_016857303.1
XM_017001815.1 569 Intron XP_016857304.1
XM_017001816.1 569 Intron XP_016857305.1
XM_017001817.1 569 Intron XP_016857306.1
XM_017001818.1 569 Intron XP_016857307.1
XM_017001819.1 569 Intron XP_016857308.1
XM_017001820.1 569 Intron XP_016857309.1
XM_017001821.1 569 Intron XP_016857310.1
XM_017001822.1 569 Intron XP_016857311.1
XM_017001823.1 569 Intron XP_016857312.1
XM_017001824.1 569 Intron XP_016857313.1
XM_017001825.1 569 Intron XP_016857314.1
XM_017001826.1 569 Intron XP_016857315.1
XM_017001827.1 569 Intron XP_016857316.1
XM_017001828.1 569 Intron XP_016857317.1
XM_017001829.1 569 Intron XP_016857318.1
XM_017001830.1 569 Intron XP_016857319.1
XM_017001831.1 569 Intron XP_016857320.1

View Full Product Details