Product Details

SNP ID
rs6682451
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:36397000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAATAAATCTGGATAATGAAT[G/T]GGGGAAAAAAACCCCCAACCGGTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LSM10 PubMed Links

Gene Details

Gene
LSM10
Gene Name
LSM10, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032881.2 Intron NP_116270.1

View Full Product Details