Product Details

SNP ID
rs11555959
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:228394709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTCCAGGAGGGCCCGAGTCAC[C/T]GGGGAGCCAGGGTGTCCCCGGACCC
Phenotype
MIM: 607868
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6742 PubMed Links

Gene Details

Gene
MIR6742
Gene Name
microRNA 6742
There are no transcripts associated with this gene.

Gene
TRIM11
Gene Name
tripartite motif containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145214.2 1658 Missense Mutation CAG,CGG Q468R NP_660215.1
XM_011544285.2 1658 Missense Mutation CAG,CGG Q391R XP_011542587.1
XM_017002412.1 1658 Missense Mutation CAG,CGG Q467R XP_016857901.1

View Full Product Details