Product Details

SNP ID
rs7599697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238322836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTAATGCCAAAGTTGATTTCTCT[C/T]CCTGTGTAAAGATAAATAAATTATG
Phenotype
MIM: 607380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAF3IP1 PubMed Links

Gene Details

Gene
TRAF3IP1
Gene Name
TRAF3 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139490.1 Intron NP_001132962.1
NM_015650.3 Intron NP_056465.2
XM_006712414.2 Intron XP_006712477.1
XM_011510944.2 Intron XP_011509246.1
XM_011510945.2 Intron XP_011509247.1
XM_011510946.2 Intron XP_011509248.1
XM_011510947.2 Intron XP_011509249.1
XM_011510948.2 Intron XP_011509250.1
XM_011510950.2 Intron XP_011509252.1
XM_017003789.1 Intron XP_016859278.1

View Full Product Details