Product Details

SNP ID
rs12693455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:152720022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAATGGTAAACTGAATGTGCAT[A/G]AAATGCAAGCTGTCAGTCTTAAATG
Phenotype
MIM: 616495 MIM: 612941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP6 PubMed Links

Gene Details

Gene
ARL6IP6
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152522.5 Intron NP_689735.1
XM_006712326.3 Intron XP_006712389.1
Gene
PRPF40A
Gene Name
pre-mRNA processing factor 40 homolog A
There are no transcripts associated with this gene.

View Full Product Details