Product Details

SNP ID
rs7596330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:215316750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGATGCAGTTTTGCTTCCTGCTT[A/T]ATTTATTTTTATGTATTTATTTATT
Phenotype
MIM: 601731
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATIC PubMed Links

Gene Details

Gene
ATIC
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004044.6 Intron NP_004035.2
XM_017004187.1 Intron XP_016859676.1

View Full Product Details