Product Details

SNP ID
rs7599903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227174997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAGATTGTGGTGCTGCAGGAGAG[A/G]ATGGGGTGTGGTGATGGAGAAGGAG
Phenotype
MIM: 120070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL4A3 PubMed Links

Gene Details

Gene
COL4A3
Gene Name
collagen type IV alpha 3 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000091.4 Intron NP_000082.2
XM_005246277.3 Intron XP_005246334.1
XM_005246280.3 Intron XP_005246337.1
XM_006712245.3 Intron XP_006712308.1
XM_011510555.1 Intron XP_011508857.1
XM_011510556.2 Intron XP_011508858.1
XM_017003295.1 Intron XP_016858784.1

View Full Product Details