Product Details

SNP ID
rs12620711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231739080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCATTGGTCACAGCCCTGTGTA[A/G]ATAAAGGGTTGGAAAGTACCTTCAT
Phenotype
MIM: 602676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE6D PubMed Links

Gene Details

Gene
PDE6D
Gene Name
phosphodiesterase 6D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291018.1 Intron NP_001277947.1
NM_002601.3 Intron NP_002592.1
XM_011511342.1 Intron XP_011509644.1

View Full Product Details