Product Details

SNP ID
hCV42700814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122795286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCTCCTGGAGAAGCCGGGAG[A/G]CGGCGGCGGCGGCGCTGCCCGCGGC
Phenotype
MIM: 602773 MIM: 608263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRC2 PubMed Links

Gene Details

Gene
DIRC2
Gene Name
disrupted in renal carcinoma 2
There are no transcripts associated with this gene.

Gene
HSPBAP1
Gene Name
HSPB1 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details