Product Details

SNP ID
hCV42712102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132419720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAACAAGGCTGTGCAGATAAAC[G/T]GCAGCACAAATTTGGTTGCTACAAA
Phenotype
MIM: 614334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC13 PubMed Links

Gene Details

Gene
DNAJC13
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015268.3 Intron NP_056083.3

View Full Product Details