Product Details

SNP ID
rs1804758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142449463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTCCAACCAAGATACATCTGG[C/T]ATAGTAAGTTTTCATCAGTAGCTTC
Phenotype
MIM: 601215 MIM: 607994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATR PubMed Links

Gene Details

Gene
ATR
Gene Name
ATR serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184.3 7798 Missense Mutation TAC,TGC Y2634C NP_001175.2
XM_011512924.1 7798 Missense Mutation TAC,TGC Y2636C XP_011511226.1
XM_011512925.1 7798 Missense Mutation TAC,TGC Y2572C XP_011511227.1
XM_011512926.2 7798 Intron XP_011511228.1
XM_017006643.1 7798 Missense Mutation TAC,TGC Y2570C XP_016862132.1
Gene
XRN1
Gene Name
5'-3' exoribonuclease 1
There are no transcripts associated with this gene.

View Full Product Details