Product Details

SNP ID
rs7625315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150873748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTGTCACTGACTAGCATATC[G/T]GCTCCCATTCCCAAGATAAGGCAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM188B2 PubMed Links

Gene Details

Gene
FAM188B2
Gene Name
family with sequence similarity 188 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017007621.1 Intron XP_016863110.1

View Full Product Details