Product Details
- SNP ID
-
rs7670092
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73407261 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTGCCCATTGATTAGTAACCCCT[C/T]GTTTCGTCCTCCCCCAGCCACTGGC
- Phenotype
-
MIM: 103600
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALB
PubMed Links
Gene Details
- Gene
- ALB
- Gene Name
- albumin
There are no transcripts associated with this gene.
View Full Product Details