Product Details

SNP ID
rs5744561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75524309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGAACTGAGTCAGTCCCAGGTG[C/T]CTTCCTTAAATACTTGTGGGTTGAA
Phenotype
MIM: 605650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLK PubMed Links

Gene Details

Gene
POLK
Gene Name
polymerase (DNA) kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016218.2 Intron NP_057302.1
XM_005248534.4 Intron XP_005248591.1
XM_006714652.3 Intron XP_006714715.1
XM_011543463.2 Intron XP_011541765.1
XM_011543464.2 Intron XP_011541766.1
XM_011543467.2 Intron XP_011541769.1
XM_017009558.1 Intron XP_016865047.1
XM_017009559.1 Intron XP_016865048.1
XM_017009560.1 Intron XP_016865049.1
XM_017009561.1 Intron XP_016865050.1
XM_017009562.1 Intron XP_016865051.1
XM_017009563.1 Intron XP_016865052.1
XM_017009564.1 Intron XP_016865053.1

View Full Product Details